1-Methoxy-2-butanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

479756-5ML
SIAL479756-5MLEA 65.8 EUR
SIAL479756-5ML
1-Methoxy-2-butanol, Sigma-Aldrich®
1-Methoxy-2-butanol
Formel: C₅H₁₂O₂
Molecular Weight: 104,15 g/mol
MDL: MFCD00059143
CAS-Nummer: 53778-73-7
EINECS: 258-763-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR