3,5-Dimethoxybenzylbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Bromomethyl)-3,5-dimethoxybenzene

480622-5G
SIAL480622-5GEA 131 EUR
SIAL480622-5G
3,5-Dimethoxybenzylbromid, Sigma-Aldrich®
3,5-Dimethoxybenzylbromid
Formel: C₉H₁₁BrO₂
Molecular Weight: 231,0864 g/mol
MDL: MFCD01321368
CAS-Nummer: 877-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR