4-Ethinylanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Aminophenyl)acetylen, 1-Ethinyl-4-aminobenzol, (4-Aminophenyl)ethin, 1-Amino-4-ethinylbenzol

M-Clarity™ Qualitätsstufe = MQ100

481122-5G
SIAL481122-5GEA 157 EUR
SIAL481122-5G
4-Ethinylanilin, Sigma-Aldrich®
4-Ethinylanilin
Formel: C₈H₇N
Molecular Weight: 117,15 g/mol
MDL: MFCD00168824
CAS-Nummer: 14235-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR