Natriumtetraethylborat, Sigma-Aldrich®

Lieferant: Merck
Gefahr


MQ100

481483-1G 481483-5G
SIAL481483-1GEA 79 EUR
SIAL481483-1G SIAL481483-5G
Natriumtetraethylborat, Sigma-Aldrich®
Sodium tetraethylborate
Formel: C₈H₂₀BNa
Molecular Weight: 150.05 g/mol
MDL: MFCD00061547
CAS-Nummer: 15523-24-7
UN: 3393
ADR: 4.2 I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR