4-Chlor-3-(trifluormethyl)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

481785-5G
SIAL481785-5GEA 93.9 EUR
SIAL481785-5G
4-Chlor-3-(trifluormethyl)benzaldehyd, Sigma-Aldrich®
4-Chloro-3-(trifluoromethyl)benzaldehyde
Formel: C₈H₄ClF₃O
Molecular Weight: 208,57 g/mol
MDL: MFCD00040722
CAS-Nummer: 34328-46-6
EINECS: 251-940-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR