5-Fluor-2-nitrobenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

481904-5G
SIAL481904-5GEA 85.4 EUR
SIAL481904-5G
5-Fluor-2-nitrobenzoesäure, Sigma-Aldrich®
5-Fluor-2-nitrobenzoesäure
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
MDL: MFCD00055635
CAS-Nummer: 320-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR