2-Chlor-3-(trifluormethyl)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

481912-1G
SIAL481912-1GEA 29 EUR
SIAL481912-1G
2-Chlor-3-(trifluormethyl)benzaldehyd, Sigma-Aldrich®
2-Chloro-3-(trifluoromethyl)benzaldehyde
Formel: C₈H₄ClF₃O
MDL: MFCD00792414
CAS-Nummer: 93118-03-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR