(R)-(+)-1,2-Dodecandiol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

482234-500MG
SIAL482234-500MGEA 202 EUR
SIAL482234-500MG
(R)-(+)-1,2-Dodecandiol, Sigma-Aldrich®
(R)-(+)-1,2-Dodecandiol
Formel: C₁₂H₂₆O₂
Molecular Weight: 202,34 g/mol
CAS-Nummer: 85514-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR