1,4-Bis(dimethylsilyl)benzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

483192-5ML
SIAL483192-5MLEA 190 EUR
SIAL483192-5ML
1,4-Bis(dimethylsilyl)benzol, Sigma-Aldrich®
1,4-Bis(dimethylsilyl)benzol
Formel: C₁₀H₁₈Si₂
MDL: MFCD00039790
CAS-Nummer: 2488-01-9
EINECS: 219-638-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR