(R)-(+)-1,2-Decandiol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

483303-500MG
SIAL483303-500MGEA 180 EUR
SIAL483303-500MG
(R)-(+)-1,2-Decandiol, Sigma-Aldrich®
(R)-(+)-1,2-Decandiol
Formel: C₁₀H₂₂O₂
Molecular Weight: 174,28 g/mol
CAS-Nummer: 87827-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR