Scandium(III)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Scandium(III) trifluoromethanesulphonate, Scandium triflate

M-Clarity™ Qualitätsstufe = MQ100

483354-1G
SIAL483354-1GEA 87.1 EUR
SIAL483354-1G
Scandium(III)trifluormethansulfonat, Sigma-Aldrich®
Scandium(III)trifluormethansulfonat
Formel: C₃F₉O₉S₃Sc
Molecular Weight: 492,17 g/mol
MDL: MFCD00192433
CAS-Nummer: 144026-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR