Bis(trimethylstannyl)acetylen, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
483613-1G
SIAL483613-1GEA
87.6
EUR
SIAL483613-1G
Bis(trimethylstannyl)acetylen, Sigma-Aldrich®
Bis(trimethylstannyl)acetylen
Formel:
C₈H₁₈Sn₂ |
MDL:
MFCD01073798 CAS-Nummer: 2117-50-2 EINECS: 218-324-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...