Boc-D-Pro-OH, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline

M-Clarity™ Qualitätsstufe = MQ100

483818-1G
SIAL483818-1GEA 27 EUR
SIAL483818-1G
Boc-D-Pro-OH, Sigma-Aldrich®
Boc-D-Pro-OH
Formel: C₁₀H₁₇NO₄
Molecular Weight: 215,25 g/mol
MDL: MFCD00063226
CAS-Nummer: 37784-17-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR