Samarium(III)iodid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Samarium (III) iodide

M-Clarity™ Qualitätsstufe = MQ100

484032-1G
SIAL484032-1GEA 218 EUR
SIAL484032-1G
Samarium(III)iodid, Sigma-Aldrich®
Samarium triiodide
Formel: SmI₃
Molecular Weight: 531,07 g/mol
CAS-Nummer: 13813-25-7
EINECS: 237-468-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR