1,8-Diiodhexadecafluoroctan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

484997-1G
SIAL484997-1GEA 119 EUR
SIAL484997-1G
1,8-Diiodhexadecafluoroctan, Sigma-Aldrich®
1,8-Diiodohexadecafluorooctane
Formel: C₈F₁₆I₂
MDL: MFCD00236093
CAS-Nummer: 335-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR