2'-Methoxyacetanilid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(2-Methoxyphenyl)acetamide

M-Clarity™ Qualitätsstufe = MQ100

485063-100G
SIAL485063-100GEA 35.4 EUR
SIAL485063-100G
2'-Methoxyacetanilid, Sigma-Aldrich®
2'-Methoxyacetanilid
Formel: C₉H₁₁NO₂
Molecular Weight: 165,1891 g/mol
MDL: MFCD00026117
CAS-Nummer: 93-26-5
EINECS: 202-233-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR