Acetonitril-[1-13C] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

485160-500MG
SIAL485160-500MGEA 770 EUR
SIAL485160-500MG
Acetonitril-[1-13C] (99% ¹³C ), Sigma-Aldrich®
Acetonitril-[1-13C]
Formel: C₂H₃N
CAS-Nummer: 31432-55-0
UN: 1648
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR