Acetonitril-[1-13C,15N] (99% ¹³C , 98% N), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
485179-250MG
SIAL485179-250MGEA
1500
EUR
SIAL485179-250MG
Acetonitril-[1-13C,15N] (99% ¹³C , 98% N), Sigma-Aldrich®
Acetonitril-[1-13C,15N]
Formel:
C₂H₃N |
MDL:
MFCD01075385 CAS-Nummer: 14320-89-9 UN: 1648 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...