Acetonitril-[13C2] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

485217-250MG
SIAL485217-250MGEA 835 EUR
SIAL485217-250MG
Acetonitril-[13C2] (99% ¹³C ), Sigma-Aldrich®
Acetonitril-[13C2]
Formel: C₂H₃N
MDL: MFCD00083867
CAS-Nummer: 1722-25-4
UN: 1648
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR