Anilin-[Ring-13C6] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

485497-250MG
SIAL485497-250MGEA 863 EUR
SIAL485497-250MG
Anilin-[Ring-13C6] (99% ¹³C ), Sigma-Aldrich®
Anilin-[Ring-13C6]
Formel: C₆H₇N
MDL: MFCD00143617
CAS-Nummer: 100849-37-4
UN: 1547
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR