Ethylacetoacetat-[2,4-13C2] (99% ¹³C ), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
485659-500MG
SIAL485659-500MGEA
820
EUR
SIAL485659-500MG
Ethylacetoacetat-[2,4-13C2] (99% ¹³C ), Sigma-Aldrich®
Ethylacetoacetat-[2,4-13C2]
Formel:
C₆H₁₀O₃ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
77504-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...