Toluol-[Ring-D5] (98% D), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Toluene-[ring-²H₅]
M-Clarity™ Qualitätsstufe = MQ200
486213-5G
SIAL486213-5GEA
807
EUR
SIAL486213-5G
Toluol-[Ring-D5] (98% D), Sigma-Aldrich®
Toluol-[Ring-D5]
Formel:
C₆D₅CH₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084186 CAS-Nummer: 1603-99-2 UN: 1294 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...