Cyclopentanon-[2,2,5,5-D4] (98% D), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Cyclopentanone-[2,2,5,5-²H₄]
M-Clarity™ Qualitätsstufe = MQ200
486507-5G
SIAL486507-5GEA
1760
EUR
SIAL486507-5G
Cyclopentanon-[2,2,5,5-D4] (98% D), Sigma-Aldrich®
Cyclopentanon-[2,2,5,5-D4]
Formel:
C₅H₄D₄O Molecular Weight: 88.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190415 CAS-Nummer: 3997-89-5 UN: 2245 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...