1,2-Dibromethan-[13C2] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

487198-1G
SIAL487198-1GEA 1030 EUR
SIAL487198-1G
1,2-Dibromethan-[13C2] (99% ¹³C ), Sigma-Aldrich®
1,2-Dibromethan-[13C2]
Formel: C₂H₄Br₂
MDL: MFCD00142466
CAS-Nummer: 33458-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR