p-Phenylendiamin-[Ring-D4] (98% D), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
p-Phenylenediamine-[ring-²H₄]
M-Clarity™ Qualitätsstufe = MQ200
487473-1G
SIAL487473-1GEA
1510
EUR
SIAL487473-1G
p-Phenylendiamin-[Ring-D4] (98% D), Sigma-Aldrich®
p-Phenylendiamin-[Ring-D4]
Formel:
C₆H₄D₄N₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
153200-72-7 UN: 1673 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...