Essigsäureanhydrid-[1,1'-13C2] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

487813-500MG
SIAL487813-500MGEA 552 EUR
SIAL487813-500MG
Essigsäureanhydrid-[1,1'-13C2] (99% ¹³C ), Sigma-Aldrich®
Essigsäureanhydrid-[1,1'-13C2]
Formel: C₄H₆O₃
MDL: MFCD00083862
CAS-Nummer: 90980-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR