Essigsäureanhydrid-[13C4] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

487821-250MG
SIAL487821-250MGEA 1140 EUR
SIAL487821-250MG
Essigsäureanhydrid-[13C4] (99% ¹³C ), Sigma-Aldrich®
Essigsäureanhydrid-[13C4]
Formel: C₄H₆O₃
MDL: MFCD00083864
CAS-Nummer: 114510-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR