Essigsäureanhydrid-[2,2'-13C2] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

487848-250MG
SIAL487848-250MGEA 718 EUR
SIAL487848-250MG
Essigsäureanhydrid-[2,2'-13C2] (99% ¹³C ), Sigma-Aldrich®
Essigsäureanhydrid-[2,2'-13C2]
Formel: C₄H₆O₃
MDL: MFCD00083863
CAS-Nummer: 98006-45-2
UN: 1715
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR