Acetophenon-[1,2-13C2] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

487872-250MG
SIAL487872-250MGEA 587 EUR
SIAL487872-250MG
Acetophenon-[1,2-13C2] (99% ¹³C ), Sigma-Aldrich®
Acetophenon-[1,2-13C2]
Formel: C₈H₈O
MDL: MFCD00083868
CAS-Nummer: 190314-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR