Mangan(II)chlorid 0.1 mol/l in wässriger Lösung, Millipore®

Lieferant: Merck

Synonyme: Manganese dichloride

48795-1ML-F
SIAL48795-1ML-FEA 66.2 EUR
SIAL48795-1ML-F
Mangan(II)chlorid 0.1 mol/l in wässriger Lösung, Millipore®
Mangan(II)chlorid
Formel: MnCl₂
Molecular Weight: 125,84 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011114
CAS-Nummer: 7773-01-5
EINECS: 231-869-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR