Ammoniumsulfat-[15N2] (5% ¹⁵N), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ammonium-15N2 sulphate, Diazanium-15N2 sulphate, Bis(ammonium-15N) sulphate, Ammonium sulphate ((15NH4)2SO4), Ammonium sulphate-15N₂, 15N Labeled ammonium sulphate

M-Clarity™ Qualitätsstufe = MQ200

488089-250G
SIAL488089-250GEA 1280 EUR
SIAL488089-250G
Ammoniumsulfat-[15N2] (5% ¹⁵N), Sigma-Aldrich®
Ammoniumsulfat-[15N2]
Formel: H₈N₂O₄S
Molecular Weight: 134,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003392
CAS-Nummer: 43086-58-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR