Ethylacetoacetat-[3-13C] (99% ¹³C ), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
489271-500MG
SIAL489271-500MGEA
578
EUR
SIAL489271-500MG
Ethylacetoacetat-[3-13C] (99% ¹³C ), Sigma-Aldrich®
Ethylacetoacetat-[3-13C]
Formel:
C₆H₁₀O₃ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
61973-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...