N-Fmoc-glycin-[U-13C2,15N] (99% ¹³C , 98% ¹⁵N)

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

489530-500MG
SIAL489530-500MGEA 2890 EUR
SIAL489530-500MG
N-Fmoc-glycin-[U-13C2,15N] (99% ¹³C , 98% ¹⁵N)
N-Fmoc-glycin-[U-13C2,15N]
Formel: C₁₇H₁₅NO₄
MDL: MFCD01075396
CAS-Nummer: 285978-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR