3-Brombenzaldehyddiethylacetal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Brom-α,α-diethoxytoluol, 1-Brom-3-(diethoxymethyl)benzol

M-Clarity™ Qualitätsstufe = MQ100

490776-10ML
SIAL490776-10MLEA 36.8 EUR
SIAL490776-10ML
3-Brombenzaldehyddiethylacetal, Sigma-Aldrich®
3-Brombenzaldehyddiethylacetal
Formel: C₁₁H₁₅BrO₂
Molecular Weight: 259,14 g/mol
MDL: MFCD01075697
CAS-Nummer: 75148-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR