Acetonitril-[13C2,15N] (99% ¹³C , 98% N), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
491675-250MG
SIAL491675-250MGEA
1920
EUR
SIAL491675-250MG
Acetonitril-[13C2,15N] (99% ¹³C , 98% N), Sigma-Aldrich®
Acetonitril-[13C2,15N]
Formel:
C₂H₃N |
MDL:
MFCD01075387 CAS-Nummer: 14149-40-7 UN: 1648 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...