Zinn(IV)-tert-butanolat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

494135-25G 494135-5G
SIAL494135-25GEA 542 EUR
SIAL494135-25G SIAL494135-5G
Zinn(IV)-tert-butanolat, Sigma-Aldrich®
Zinn(IV)-tert-butanolat
Formel: C₁₆H₃₆O₄Sn
MDL: MFCD00156551
CAS-Nummer: 36809-75-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR