4-Hydroxy-3-nitropyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Nitro-4-pyridinol

M-Clarity™ Qualitätsstufe = MQ200

495212-5G
SIAL495212-5GEA 55.9 EUR
SIAL495212-5G
4-Hydroxy-3-nitropyridin, Sigma-Aldrich®
4-Hydroxy-3-nitropyridine
Formel: C₅H₄N₂O₃
Molecular Weight: 140,10 g/mol
MDL: MFCD00128874
CAS-Nummer: 5435-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR