Natriumcyanamidat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Sodium hydrogencyanamide
M-Clarity™ Qualitätsstufe = MQ100
495905-25G
495905-5G
SIAL495905-25GEA
169
EUR
SIAL495905-25G
SIAL495905-5G
Natriumcyanamidat, Sigma-Aldrich®
Natriumcyanamidat
Formel:
CHN₂Na Molecular Weight: 64.02 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00066500 CAS-Nummer: 17292-62-5 EINECS: 241-322-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...