Ammoniumcobalt(II)sulfat Hexahydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Cobalt(II)ammoniumsulfat Hexahydrat
M-Clarity™ Qualitätsstufe = MQ100
497363-25G
SIAL497363-25GEA
66.4
EUR
SIAL497363-25G
Ammoniumcobalt(II)sulfat Hexahydrat, Sigma-Aldrich®
Ammoniumcobalt(II)sulfat Hexahydrat
Formel:
H₈CoN₂O₈S₂·6H₂O Dichte: 1,902 g/cm³ (25 °C) |
MDL:
MFCD00151049 CAS-Nummer: 13586-38-4 EINECS: 237-043-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...