3-Brom-5-chlor-2-hydroxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Bromo-5-chlorosalicylaldehyde

M-Clarity™ Qualitätsstufe = MQ100

497592-5G
SIAL497592-5GEA 116 EUR
SIAL497592-5G
3-Brom-5-chlor-2-hydroxybenzaldehyd, Sigma-Aldrich®
3-Bromo-5-chloro-2-hydroxybenzaldehyde
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
MDL: MFCD00051690
CAS-Nummer: 19652-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR