2-Chlor-5-iodpyridin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
498181-2G
SIAL498181-2GEA
117
EUR
SIAL498181-2G
2-Chlor-5-iodpyridin, Sigma-Aldrich®
2-Chlor-5-iodpyridin
Formel:
C₅H₃ClIN Molecular Weight: 239.44 g/mol |
MDL:
MFCD01863635 CAS-Nummer: 69045-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...