2-Acetyl-4-methylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(4-Methylpyridin-2-yl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

499234-1G
SIAL499234-1GEA 74.7 EUR
SIAL499234-1G
2-Acetyl-4-methylpyridin, Sigma-Aldrich®
2-Acetyl-4-methylpyridin
Formel: C₈H₉NO
Molecular Weight: 135,1632 g/mol
MDL: MFCD01863638
CAS-Nummer: 59576-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR