Benzo[b]furan-2-boronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

499943-5G
SIAL499943-5GEA 91.3 EUR
SIAL499943-5G
Benzo[b]furan-2-boronsäure, Sigma-Aldrich®
Benzo[b]furan-2-boronsäure
Formel: C₈H₇BO₃
MDL: MFCD00236019
CAS-Nummer: 98437-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR