di-Natrium-β-glycerophosphat Pentahydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: ortho-Phosphorsäure(2-hydroxy-1-(hydroxymethyl)ethyl)ester Dinatriumsalz Pentahydrat

50020-500G
SIAL50020-500GEA 570 EUR
SIAL50020-500G
di-Natrium-β-glycerophosphat Pentahydrat, Sigma-Aldrich®
di-Natrium-β-glycerophosphat Pentahydrat
Formel: C₃H₇Na₂O₆P·5H₂O
Molecular Weight: 306,11 g/mol
MDL: MFCD00002135
CAS-Nummer: 13408-09-8
EINECS: 212-464-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR