1-Butyl-1-methylpyrrolidiniumdicyanamid, Sigma-Aldrich®

Lieferant: Merck

50893-50G-F
SIAL50893-50G-FEA 691 EUR
SIAL50893-50G-F
1-Butyl-1-methylpyrrolidiniumdicyanamid, Sigma-Aldrich®
1-Butyl-1-methylpyrrolidiniumdicyanamide
Formel: C₁₁H₂₀N₄
MDL: MFCD08458923
CAS-Nummer: 370865-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR