Trihexyl(tetradecyl)phosphoniumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
Lieferant: Merck
50971-50G-F
50971-5G-F
SIAL50971-50G-FEA
260
EUR
SIAL50971-50G-F
SIAL50971-5G-F
Trihexyl(tetradecyl)phosphoniumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
Trihexyl(tetradecyl)phosphoniumbis(trifluormethylsulfonyl)imid
Formel:
C₃₄H₆₈F₆NO₄PS₂ |
MDL:
MFCD04117695 CAS-Nummer: 460092-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...