(R)-3-(Boc-amino)-3-(4-fluorphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-3-(4-fluorophenyl)propanoic acid

51006-500MG-F
SIAL51006-500MG-FEA 340 EUR
SIAL51006-500MG-F
(R)-3-(Boc-amino)-3-(4-fluorphenyl)propionsäure, Sigma-Aldrich®
(R)-3-(Boc-amino)-3-(4-fluorphenyl)propionsäure
Formel: C₁₄H₁₈FNO₄
Molecular Weight: 283,2954 g/mol
MDL: MFCD03427957
CAS-Nummer: 479064-94-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR