4-(Trifluormethoxy)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dihydroxy(4-(trifluormethoxy)phenyl)boran, 4-(Trifluormethoxy)benzolboronsäure, (4-(Trifluormethoxy)phenyl)metaborsäure

510130-5G
SIAL510130-5GEA 80.2 EUR
SIAL510130-5G
4-(Trifluormethoxy)phenylboronsäure, Sigma-Aldrich®
4-(Trifluormethoxy)phenylboronsäure
Formel: C₇H₆BF₃O₃
Molecular Weight: 205,93 g/mol
MDL: MFCD01074648
CAS-Nummer: 139301-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR