Methyltrifluorpyruvat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3,3,3-trifluoropyruvate, Methyl 3,3,3-trifluoro-2-oxopropanoate

M-Clarity™ Qualitätsstufe = MQ100

510246-5G
SIAL510246-5GEA 36.3 EUR
SIAL510246-5G
Methyltrifluorpyruvat, Sigma-Aldrich®
Methyltrifluorpyruvat
Formel: C₄H₃F₃O₃
Molecular Weight: 156,06 g/mol
MDL: MFCD00114936
CAS-Nummer: 13089-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR