Ethyltrifluorpyruvat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 3,3,3-trifluoro-2-oxopropanoate, Ethyl 3,3,3-trifluoropyruvate

M-Clarity™ Qualitätsstufe = MQ100

510254-5G
SIAL510254-5GEA 21.2 EUR
SIAL510254-5G
Ethyltrifluorpyruvat, Sigma-Aldrich®
Ethyltrifluorpyruvat
Formel: C₅H₅F₃O₃
Molecular Weight: 170,09 g/mol
MDL: MFCD00114935
CAS-Nummer: 13081-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR